Digital Timer
Supplier: Taylor Precision Products
5806
470093-772CS 124.25 CAD
470093-772
Digital Timer
Timers Digital Timers
The multi-purpose digital timer is versatile and easy to use.
  • Small and compact design
  • Counts up and counts down
  • Clip/magnet/stand positioning
This lightweight timer can count up (stopwatch function) or count down. A 0.7 LCD readout provides a clear display that can be read from a distance. The timer is capable of programming up to 99 minutes.
Power supply: AAA battery
Packaging: Retail card.