Benedict's Qualitative Solution

Supplier: ALDON CORP SE
BB0090-100ML BB0090-1L BB0090-3.8L BB0090-500ML
470300-360EA 8.25 CAD
470300-360 470300-358 470300-364 470300-362
Benedict's Qualitative Solution
Chemicals for Science Education
CAS Number: Mixture
Formula Weight: Mixture
Density: 1.0 g/mL
Boiling and Freezing Point: 100°C, 0°C
Solubility: Water
Synonyms: Benedict's Sugar Test Reagent, Benedict's Solution
Shelf Life: 12 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR