Printer for BioClave Benchtop Autoclaves
Supplier: Ward's
B4000-M-E B4000-16-E
470230-602EA 7924.8 CAD
470230-602 470230-604
Printer for BioClave Benchtop Autoclaves
Sterilizers Autoclaves (Steam Sterilizers)
The benchtop autoclave's steam sterilizes research tools and labware.
  • Rapid, complete sterilization for educational/research use
  • Built-in water tank doesn't require external water supply
  • Safety interlocks prevent door from opening while pressurized
  • Stainless steel sterilization chambers
  • One-year warranty
BioClave Mini™: The 8 liter BioClave Mini is preprogrammed to meet basic sterilization needs. Temperature can be selected at either 121°C or 134°C, and the sterilization time is fully adjustable.
BioClave 16™: The 16 liter BioClave offers preset cycle options (standard programs), for use with liquids, wrapped research instruments or unwrapped instruments/plasticware. An additional option “Dry Only” is also available for additional drying time to be added to the end of a cycle. For special applications, requiring variations in cycle parameters,all settings on the BioClave 16 are fully adjustable.
BioClave 28™: Features an increased chamber size with a diameter of 32cm. With this larger internal chamber, the BioClave 28 is our only model capable of sterilizing 1 and 2 liters flasks (and bottles). Despite the increased chamber size, the unit is still relatively compact, suitable for benchtop use.
Ordering information: One year warranty.
Caution: Not intended for medical use or with food.