Ward's® Basic Microbiological Techniques Lab Activity
Supplier: Ward's
470030-196KT 521.39 CAD
470030-196
Ward's® Basic Microbiological Techniques Lab Activity
Educational Materials Biology Educational Materials Intro to Science Learning Activities
Learn How to Identify Bacteria Using Several Different Staining Procedures
  • Demonstrates multiple techniques
  • Includes two bacterial cultures
  • Materials for six setups
  • Time Required: ~ 90 minutes
By employing culturing and staining techniques including sterile transfer, streak plating, Gram staining, and endospore staining, students identify the included “unknown” bacteria. These procedures also familiarize students with sterile technique and preparation of microscope slides. Tube cultures: Bacillus cereus and Escherichia coli. Includes a teacher’s guide, and student copymaster.
Canada: Containment Level 2 – PHAC License required.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.