Barium Nitrate

Supplier: ALDON CORP SE

BB0055-CAPS BB0055-CAP BB0061-500ML BB0055-100G BB0055-500G BB0055-100GR BB0055-500GR
470300-336EA 116.55 CAD
470300-336 470300-338 470300-340 470300-342 470300-344 470300-346 470300-348
Barium Nitrate
Chemicals for Science Education

CAS Number: 10022-31-8
Formula Weight: 261.35
Formula: Ba(NO3)2
Hazard Info: Oxidizer, Toxic
Density (g/mL): 3.24
Boiling Point (°C): Decomposes
Freezing Point (°C): 592
Solubility: Water
Synonyms: Barium Dinitrate, Nitric Acid Barium Salt
Shelf Life (months): 36
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR