Barium Hydroxide Octahydrate

Supplier: ALDON CORP SE

BB0050-100G BB0050-500G BB0045-500G BB0043-500ML BB0045-100GR BB0048-500ML
470045-584EA 29.4 CAD
470045-584 470300-328 470300-332 470300-324 470300-330 470225-772
Barium Hydroxide Octahydrate
Chemicals for Science Education

CAS Number: 12230-71-6
Formula Weight: 315.48
Formula: BaOH2·8H2O
Density: 2.18 gg/mL
Solubility: Water, Alcohol and Ether
Synonyms: Barium Hydrate, Barium Octahydrate
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is toxic.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR