Barfoed Reagent

Supplier: ALDON CORP SE

BB0025-500ML BB0025-100MLR
470300-300EA 58.95 CAD
470300-300 470300-298
Barfoed Reagent
Chemicals for Science Education

CAS Number: Mixture
Formula Weight: Mixture
Density: Approximately 1 g/mL
Boiling Point and Freezing Point: Approximately 100 °C, Approximately 0°C
Solubility: Water
Synonyms: Glucose Test Solution
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR