Bare Nichrome Wire
Supplier: Arcor Electronics
4 OZ D24 D26 D20 D-28
470005-952EA 60.2 CAD
470005-952 470200-354 470200-356 470201-280 470201-282
Bare Nichrome Wire
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Bare Nichrome Wire can handle temperatures of up to 1010°C (1850°F).
  • Alloy of Nickel and Chromium
  • Grams: 113
  • Wire Can Handle Temperatures up to 1010°C (1850°F)
  • High Performance
  • Ideal for Rheostats and Controls Requiring Compact Resistance
It's particularly suited to heavy duty rheostats and controls that require compact resistance.
The wire's alloy is made of nickel and chromium.