Balsam Canada

Supplier: ALDON CORP SE

BB0005-100ML BB0005-25ML BB0001-30ML BB0020-25ML
470300-290EA 288.65 CAD
470300-290 470300-292 470300-294 470300-296
Balsam Canada
Chemicals for Science Education

CAS Number: Mixture
Formula Weight: Mixture
Formula: Mixture
Density (g/mL): 0.87
Boiling Point (°C): 139-141
Synonyms: Synthetic Mounting Medium, Piccolyte Slide Mounting Media
Shelf Life (months): 36
Storage: Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR