Ballistics Car
Supplier: VWR International
PH0355APL
470015-994EA 116.3 CAD
470015-994
Ballistics Car
Educational Materials Physics Educational Materials Mechanics Learning Activities
The Ballistics Car demonstrates an integral law of motion.
  • Steel Ball Provided
  • Release Pin Provided
  • Cord Provided
  • Car Demonstrates Integral Law of Motion
  • Barrel Permits Two Heights of Ball Flight
At first explanation, this demonstration seems impossible. However, when students watch this Ballistics Car in action, they will believe and comprehend an important physical science concept.
Simply push the car and eject the steel ball from the attached, spring-loaded barrel. The ball falls back down into the barrel of the moving car, illustrating that the forward motion of the ball is the same as that of the vehicle from which it was ejected. The barrel permits two heights of ball flight.
Ordering information: The kit includes a steel ball, release pin, and cord.