Ward's® Bacti-Spreader
Supplier: Ward's
151993
470004-860EA 9.95 CAD
470004-860
Ward's® Bacti-Spreader
Spreaders Cell Spreaders
Special Design Evenly Distributes Bacteria
  • Reusable for multiple inoculations
The stainless steel rod has been specifically designed for greater contact with agar for more even distribution of bacteria.