Archimedes’ Block Model
Supplier: PRECISION MOULDED POLYMERS LTD
A-M9024
470014-938EA 71.65 CAD
470014-938
Archimedes’ Block Model
Educational Materials Physics Educational Materials Mechanics Learning Activities
Your Students Will Shout Eureka When They Use this Simple Apparatus.
  • Includes block, weights, spring scale, and a teacher’s guide
  • Size: 2"W x 1 1/2"H x 2"D
A graduated block, water, and a beaker you provide are all you need to re-create Archimedes’ discovery that a body immersed in a fluid becomes lighter by an amount equal to that of the fluid it displaces. Add the included weights to the cube to demonstrate displacement; add enough to submerge the block and measure its mass with a spring scale to illustrate the effects of buoyancy. You can also teach the concepts of specific gravity and density.