Ward's® Physiology of the Circulatory System Lab Activity
Supplier: Avantor
470009-226KT 100.95 CAD
470009-226 470194-706
Ward's® Physiology of the Circulatory System Lab Activity
Educational Materials Biology Educational Materials AP Biology Learning Activities
View Daphnia Heart Rate Easily
  • Teaches technique for measuring blood pressure
  • Comparative physiology featuring Daphnia
  • Specifically designed viewing chambers for viewing Daphnia
  • Time Required: 75 minutes
Students learn how to measure blood pressure, pulse, and heart rate, and assess the effects that varying physiological conditions have on their rates, in this practical, comprehensive lab activity. For comparison, students also study the heart rate of Daphnia, varying the temperature and viewing the subsequent changes that take place in theMore Product Information