Aniline Blue

Supplier: ALDON CORP SE

AA0360-25G AA0361-25ML
470300-274EA 84.35 CAD
470300-274 470300-276
Aniline Blue
Chemicals for Science Education

CAS Number: 28631-66-5
Formula Weight: 737.73
Formula: C32H25N3Na2O9S3
Hazard Info: Irritant
Boiling Point (°C): Decomposes
Synonyms: Acid Blue 22, C.I. No. 42755
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR