Analog Stopwatch
Supplier: VWR International
PH0358A
470123-440EA 314.45 CAD
470123-440
Analog Stopwatch
Timers Analogue Stopwatches
Count up to 29:59 with resolution of ¹/₅ sec. It is nickel-plated and features two-dials and a hard carrying case.
Size: 2"Ø.