Ammonium Thiocyanate

Supplier: ALDON CORP SE

AA0341-500ML AA0340-100G AA0340-500G AA0340-100GR AA0340-500GR
470300-258EA 32.55 CAD
470300-258 470300-260 470300-262 470300-264 470300-266
Ammonium Thiocyanate
Chemicals for Science Education

CAS Number: 1762-95-4
Formula: NH4SCN
Density: 1.3 g/mL
Freezing Point: Approximately 149°C
Solubility: Water, Alcohol, Acetone, and Ammonia
Synonyms: Thiocyanic Acid Ammonium Salt
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: The product is toxic.

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR