Ammonium Persulfate
Supplier: ALDON CORP SE
CAS Number: 7727-54-0
Formula Weight: 228.20
Formula: (NH4)2S2O8
Hazard Info: Oxidizer, Reactive
Density (g/mL): 1.92
Solubility: Water
Synonyms: Ammonium Peroxydisulfate, Diammonium Peroxydisulfate
Shelf Life (months): 36
Storage: Yellow
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...