Ammonium Nitrate

Supplier: ALDON CORP SE

AA0303-500ML AA0301-500ML AA0302-CONC AA0295-100GR AA0295-500G
470300-224EA 25.6 CAD
470300-224 470300-226 470300-228 470300-230 470300-232 470300-234 470300-236
Ammonium Nitrate
Chemicals for Science Education

CAS Number: 6484-52-2
Formula: NH4NO3
Density: 1.73 g/mL
Boiling and Freezing Point: Decomposes, 169 °C
Solubility: Water, Alcohol, and Alkalies
Synonyms: Nitric Acid Ammonium Salt
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: The product is a toxic, reactive irritant.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR