Ammonium Hydroxide

Supplier: ALDON CORP SE

AA0275-1L AA0275-500ML AA0273-30ML AA0273-500ML AA0264-500MLR AA0265-3.8L AA0264B-2.5LR AA0280-500ML AA0274-500ML AD-AA0264P-2.5L AD-AA0264P-PK/6
470300-204EA 23.45 CAD
470300-204 470300-206 470300-208 470300-210 470300-212 470300-216 470300-214 470225-686 470225-764 470354-374 470354-384
Ammonium Hydroxide
Chemicals for Science Education

CAS Number: 1336-21-6
Formula: NH4OH
Density: 0.900 g/mL
Boiling and Freezing Point: 36°C, -77°C
Synonyms: Ammonium Hydroxide Water Solution
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: Corrosive, irritant and toxic.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR