Ammonium Dichromate

Supplier: ALDON CORP SE

AA0255-100G AA0255-500G AA0255-500GR
470300-198EA 25.45 CAD
470300-198 470300-200 470300-202
Ammonium Dichromate
Chemicals for Science Education

CAS Number: 7789-09-5
Formula Weight: 252.06
Formula: (NH4)2Cr2O7
Hazard Info: Oxidizer, Flammable, Toxic, Irritant
Density (g/mL): 2.15
Freezing Point (°C): 170
Solubility: Water and Alcohol
Synonyms: Ammonium Bichromate, Dichromic Acid Diammonium Salt, Diammonium Dichromate(VI)
Shelf Life (months): 12
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR