Amber Latex Tubing
Supplier: Primeline
026RA 034RA
62996-786RL 184.35 CAD
62996-786 62996-788
Amber Latex Tubing
Tubing Transfer Tubing
Medium wall sulfur-free tubing.
  • Smooth finish
  • Translucent
This tubing is recommended for gas analysis or other applications where contamination must be avoided. Its clinging characteristics make it excellent for glass connections.