Amber Glass Dropping Bottles
Supplier: EMPIRE/EMCO
01A002A W/ 01A010A 405-5D-24-GR
470188-926EA 4.65 CAD
470188-926 470188-928 470189-418
Amber Glass Dropping Bottles
Bottles Dropping Bottles
Ideal for Small Quantities of Light Sensitive Chemicals.
  • Store stock reagents and solutions
  • Matching pipet assemblies are available separately