Aluminum Sulfate 18-Hydrate

Supplier: ALDON CORP SE

AA0201-500G AA0201-500GR AA0202-500ML AA0204-CONC
470300-160EA 35.65 CAD
470300-160 470300-162 470300-164 470300-166
Aluminum Sulfate 18-Hydrate
Chemicals for Science Education

CAS Number: 7784-31-8
Formula: Al2(SO4)3·18H2O
Density: 1.61 g/mL
Boiling Point: Decomposes
Freezing Point : 86°C
Solubility: Water
Shelf Life: 36 months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR