Aluminum Oxide
Supplier: ALDON CORP SE
CAS Number: 1344-28-1
Formula Weight: 101.96
Formula: Al2O3
Hazard Info: Irritant, Nuisance
Density (g/mL): 3.5
Boiling Point (°C): 2980
Freezing Point (°C): 2050
Solubility: Insoluble in Water, Low solubility in Acids
Synonyms: Alumina, Aluminum Trioxide
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...