Aluminum Nitrate 9-Hydrate

Supplier: ALDON CORP SE

AA0184-500G AA0184-100GR AA0184-500GR AA0183-500ML AA0182-CONC
470300-138EA 105.6 CAD
470300-138 470300-140 470300-142 470300-144 470300-146
Aluminum Nitrate 9-Hydrate
Chemicals for Science Education

CAS Number: 7784-27-2
Formula Weight: 375.13
Formula: Al(NO3)3·9H2O
Hazard Info: Oxidizer, Toxic
Freezing Point (°C): 70
Solubility: Cold Water, Decomposes in Alcohol and Acetone
Synonyms: Aluminum Trinitrate, Aluminum Nitrate Nonahydrate
Shelf Life (months): 36
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR