Aluminum Chloride Hexahydrate

Supplier: ALDON CORP SE

AA0165-500GR AA0169-500ML
470300-120EA 89.65 CAD
470300-120 470300-124
Aluminum Chloride Hexahydrate
Chemicals for Science Education

CAS Number: 7784-13-6
Formula Weight: 241.43
Formula: AlCl3·6H2O
Hazard Info: Irritant, Corrosive
Density (g/mL): 2.398-2.440
Solubility: Water and Alcohol
Shelf Life (months): 12
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR