Aluminum

Supplier: VWR International
AA0140-30G P50070/4 AA0141-500G AA0135-100G DPK9202535 AA0135-500G AA0145-500G AA0140-30G AA0145-100G
470149-542CS 281.65 CAD
470149-542 470174-014 470300-128 470163-744 470300-126 470300-136 470300-168 470300-134 470300-132
Aluminum
Chemicals for Science Education
CAS Number: 7429-90-5
Formula: Al
Density (g): 2.7 g/mL
Shelf Life: 36 Months
Storage: Green/Red

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR