Alloy Tuning Fork Sets
Supplier: VWR International
P36135
470148-640CS 622.6 CAD
470148-640 470104-594
Alloy Tuning Fork Sets
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Two sets of popular tuning forks are available.
  • Useful Selection of Frequencies
  • Two Sets Available
  • Setting from A to G
One set features four tuning forks (256 C, 320 E, 384 G, 512C).
The other set features eight tuning forks (256 C, 320 E, 384 G, 480 B, 288 D, 341.3 F, 426.7 A, 512 C).