Alligator Test Clips
Supplier: TOP LAB CORP.
CTA-004
470176-120EA 2.85 CAD
470176-120
Alligator Test Clips
Electrical Sockets
Long nosed clips specially suitable for electrical connections.
  • Constructed of plated steel
They have a 5 A capacity and a jaw spread of 9.5 mm.
Packaging: Pack of 12.