All Glass Aquaria
Supplier: Pet World Inc
10100 10290
470005-118EA 53.95 CAD
470005-118 470005-120 470005-122 470005-124 470005-128 470021-448
All Glass Aquaria
Specimens Live Specimens Aquaria - Living Material
With no metal corners or panels, these plate glass aquaria allow students to carefully observe aquatic life from all angles.
  • Non-Metal Construction
  • Corner Seals Resist Cracking When Dry
  • Easy-To-Clean and Store
In addition, the absence of metal components means that these aquaria can be used as saltwater tanks without corrosion. The glass sides are permanently joined with an invisible, leakproof cement, and the glass bottom is set in attractive, high-impact plastic molding for added protection.
All top edges are protected against chipping by vinyl molding. In addition, these aquaria can be left empty for any length of time without developing leaks.