Alizarin Yellow R
Supplier: ALDON CORP SE
CAS Number: 1718-34-9
Formula Weight: 309.22
Formula: C13H8N3O5Na
Hazard Info: Irritant
Density (g/mL): >1
Boiling Point (°C): Decomposes
Freezing Point (°C): 250
Solubility: Many Organic Solvents, Hot Methyl Alcohol and Water
Synonyms: Mordant Yellow 3R, C.I. No. 14095
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...