Alizarin Red S

Supplier: ALDON CORP SE

AA0103-100ML AA0100-10G
470300-106EA 25.35 CAD
470300-106 470300-108
Alizarin Red S
Chemicals for Science Education

CAS Number: 130-22-3
Formula Weight: 342.28
Formula: C14H7NaO7S
Hazard Info: Irritant
Synonyms: Mordant Red 3, C.I. No. 58005
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR