Ward's® Live Alcaligenes faecalis Culture

Supplier: Avantor
8750
470176-382EA 55.3 CAD
470176-382 470179-186
Ward's® Live Alcaligenes faecalis Culture
Specimens Live Specimens Bacteria - Living Material
A Common Intestinal Flora Isolate, Demonstrates Alpha Hemolysis on Blood Agar Growth Medium

  • Biosafety Level: 1
  • Canada: Containment Level 2 - PHAC Licence required
  • Gram Stain: Negative
  • Cell Morphology: Cocci or coccal-rods
  • Motility: Positive
  • Oxidase: Positive
  • Respiration: Aerobic
  • Spores: Yes - subterminally located, oval in shape.
  • Hemolysis: Alpha on blood agar

All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.

This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.

Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.

Bacterial and fungal cultures are restricted and can only be ordered by educational and research professionals.

Ward's Science is not affiliated with the American Type Culture Collection and does not provide corresponding ATCC numbers for our organisms.

Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR