Agrobacterium tumafaciens
Supplier: VWR
WARD470179-044
470179-044EA 22.05 CAD
470179-044
Agrobacterium tumafaciens
Specimens Live Specimens Bacteria - Living Material
A. tumefaciens is a pathogenic soil bacterium known to cause grown gall disease, in nut trees and the plants of some stone fruits.
  • Biosafety Level: 1
  • Gram Stain: Negative
  • Cell Morphology: Rods
  • Oxidase: Positive
  • Indole: Negative
  • Respiration: Aerobic
A. tumefaciens is also known as Rhizobium radiobacter, and Agrobacterium radiobacter.
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: This organism is pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.