Agarose

Supplier: ALDON CORP SE

AA0083-25G AA0083-5G AA0083-125G
470300-098EA 131.2 CAD
470300-098 470300-100 470300-096
Agarose
Chemicals for Science Education

CAS Number: 9012-36-6
Density (g/mL): >1
Freezing Point (°C): 260
Solubility: Boiling Water
Synonyms: Agarose Low EEO
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR