Agar

Supplier: ALDON CORP SE

AA0075-25G AA0075-500G AA0075-100G
470300-092EA 23 CAD
470300-092 470300-094 470300-090
Agar
Chemicals for Science Education

CAS Number: 9012-36-6
Density: 1 g/mL
Freezing Point: 260 °C
Shelf Life: 36 Months
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR