A-Frame Base
Supplier: Wards
CH0655-THRED CH0655-LG
470121-238EA 80.35 CAD
470121-238 470121-256
A-Frame Base
Retort Stands
This cast iron base features threads for rods sized 1/2 - 13.
Legs are 21 cm long on the small base and 25 cm long on the large base.