Advanced Enzymology Lab Activity
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
8-130
470006-272EA 1226.95 CAD
470006-272
Advanced Enzymology Lab Activity
Educational Materials Biology Educational Materials Cell Biology Learning Activities
Virtually a Complete Mini-Course in Itself
The many topics included in this activity kit include the effects of substrates, pH, temperature, and inhibitors on the reaction rate. It comes with enough materials for 12 setups, an instructor’s manual, and student data sheets.
Ordering information: This item contains a coupon for perishable materials that must be redeemed from the manufacturer before use. Please allow sufficient time to account for redemption. These materials are shipped from the supplier via 2nd day delivery on Mondays, Tuesdays, or Wednesdays only and must be received promptly.