Adjustable Bunsen Burners
Supplier: Shiv Dial Sud & Sons
C-1482
470149-518CS 767.65 CAD
470149-518
Adjustable Bunsen Burners
Heaters Gas Burners
Precise Flame Control.
  • Gas Inlet: 5/16" O.D.
  • Mixing Tube: 1/2" O.D.
  • Height: 63/8"
These are similar to the standard burner, but with a fine pitch needle valve for flame adjustment and aluminum mixing tube with flame stabilizer.