ADA Instructor’s Desk

Supplier: Diversified Woodcrafts
1214K-L-ADA 1216K-L-ADA
470117-450EA 9264.15 CAD
470117-450 470117-452
ADA Instructor’s Desk
Furniture Tables and Workbenches
Wheelchair accessible instructor's desk with large knee space.

  • ¾" phenolic or epoxy resin surface
  • Constructed of solid oak and oak veneers
  • Chemical resistant, earth-friendly UV finish
  • Dovetailed drawers are operated smoothly on epoxy coated steel glides with nylon bearings
  • GFI protected AC duplex receptacle

This unit has a roll-under sink with paddle handle faucet and gas fixture and also includes a greenlaw arm.

Dimensions: 14W×10D×5"H, sink; 19D×27H×30"W, interior seating clearance

Certifications: SEFA compliant and MAS Certified Green. Meets ADA requirements.

Ordering information: Choose from either a 1" solid epoxy or phenolic resin top with an epoxy sink and a rod socket.

Delivery information: Includes a duplex outlet, file drawer, three smaller drawers and a pull-out writing board.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR