Fischer Technical Acrylic Pump Plate
Supplier: FISCHER TECHNICAL CO SE
AP-001
470121-116EA 409.15 CAD
470121-116
Fischer Technical Acrylic Pump Plate
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
The durable acrylic pump plate accommodates bell jars up to 9" Ø.
  • Durable
  • Vacuum release valve included
  • Rubber gasket included
  • Barbed-hose connection included
  • Plate fits bell jars up to 9" Ø.
It includes a rubber gasket for a tight seal.
The plate also includes a vacuum release valve and 3/8" barbed-hose connection.