Acrylic Buret
Supplier: United Scientific Supplies
11503 470148-702
470111-676EA 181.35 CAD
470111-676 470148-702
Acrylic Buret
Burettes
These hardy burets are ideal for classroom use.
  • Acrylic
  • Transparent
  • Leakproof Stopcock Included
  • Burets Unaffected by Most Dilute Aqueous Solutions
  • Heavy-Duty
The transparent, acrylic burets are supplied with self-lubricating, leakproof PMP stopcock.
They are unaffected by most dilute aqueous solutions except ammonia and HF and are not recommended for use with ethanol or organic solvents.
Order Now
SPECIFICATIONS
Environmentally Preferable