Aceto-Orcein Stain

Supplier: ALDON CORP SE

AA0030-100ML AA0030-30ML
470300-072EA 80.5 CAD
470300-072 470300-074
Aceto-Orcein Stain
Chemicals for Science Education

CAS Number: Mixture
Formula Weight: Mixture
Formula: Mixture
Density (g/mL): 1.049
Boiling Point (°C): 118.1
Freezing Point (°C): 16.7
Synonyms: Aceto-Orcein Biological Stain
Shelf Life (months): 36
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR