Acetic Acid

Supplier: ALDON CORP SE

AA0005-250ML AA0008-2.5LR AA0008-500MLR AA0005-100MLR AA0005-2.5L AA0005-PK/2R AA0010-1L AA0007-500ML AA0005-CONC AA0015-1L AA0005-500MLR AD-AA0009-3.8L AA0003-500ML AA0004-200ML AD-AA0005P-2.5L AD-AA0005P-PK/6
470300-028EA 51.75 CAD
470300-028 470300-030 470300-032 470300-034 470300-036 470300-040 470300-042 470300-044 470300-046 470300-048 470300-038 470219-202 470225-684 470225-820 470354-376 470354-378
Acetic Acid
Chemicals for Science Education

CAS Number: 64-19-7
Formula: CH3COOH
Density: 1.049 g/ml
Boiling Point: 118.1°C
Freezing Point: 16.7°C


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: The item is corrosive, flammable and toxic.

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR