2,3,5-Triphenyl-2H-Tetrazolium Chloride

Supplier: ALDON CORP SE

TT0270-25G TT0270-5G
470300-010EA 167.7 CAD
470300-010 470300-012
2,3,5-Triphenyl-2H-Tetrazolium Chloride
Chemicals for Science Education

CAS Number: 298-96-4
Formula Weight: 334.81
Formula: C19H15CIN4
Boiling Point (°C): Decomposes
Freezing Point (°C): 250
Solubility: Water and Alcohol
Synonyms: Triphenyltetrazolium Chloride
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR