Boreal2 Microscopes, EM Series
Supplier: VWR
SK51 SK52 SK54
470014-504EA 361.05 CAD
470014-504 470014-506 470014-508 470014-510
Boreal2 Microscopes, EM Series
Microscopes
One of our easiest-to-use models, this is the perfect introductory microscope for students.
  • Full-Size Optics
  • Ergonomic Body Style
  • Features Designed to Prevent Tampering
  • Objectives: 4x, 10x, and 40x
  • Boreal Lifetime Warranty
It boasts a gliding stage with stage clips for one-hand use, focus dots that make focusing quick and easy, and color-coded and distinctly labeled objectives.
The microscope also features an all-metal chassis, illumination without heat through a bright, white LED, and up to 400x magnification.
Ordering information: This product includes a manufacturer's warranty.