Boreal2 Microscopes, HM Series
Supplier: VWR
SK102 SK110 SK101
470014-512EA 570.15 CAD
470014-512 470014-514 470016-038
Boreal2 Microscopes, HM Series
Microscopes
With all-metal construction, a rugged, student-proof design, and precision optics, Boreal2 HM Microscopes are ideal for middle and high school students.
  • Rotatable, 45°-Inclined Viewing Head
  • Parfocal and Parcentered Optics
  • Lifetime Manufacturer's Warranty
  • Designed for Middle and High School Students
  • Total Magnification of 400x
A bit more advanced than the EM series, this microscope is designed for middle and high school students. Features include a gliding stage with one-touch stage clips, recessed eyepiece screws to prevent the disassembly of the microscope, a removable cord holder for easy storage, tension control to halt stage drift, large and easy-to-read magnification labels, and a reversed nose piece to protect objectives.
This microscope also features both a coarse and fine focus, has bright, white LED lighting, and offers a total magnification of 400x.
Ordering information: This product includes a manufacturer's warranty.