Boreal2 Microscopes, HM Advanced Series
Supplier: VWR
SK121 SK122 SK132 SK123BINOA
470014-518EA 826.8 CAD
470014-518 470014-520 470014-528 470018-032
Boreal2 Microscopes, HM Advanced Series
Microscopes
Perform quantitative experiments over a wide frequency range with these microscopes.
  • Objectives: 4x, 10x, 40x, and 100x
  • Large Magnification Labels
  • Protection against Disassembly
  • Tension Control
  • Charger Included
Bright, white, cold LED lighting and coated-glass optics give excellent images. The 4x, 10x, 40x, and 100x objectives have a wide color band with large, easy-to-read magnification labels on two sides for simplified viewing. Full-size, stable bodies hold a host of student-proof features, including locked-on stage clips, eye pieces, and objectives that offer protection against disassembly.
A four-hole reversed nosepiece protects objectives, and tension control stops stage drift. The microscopes are available in corded or cordless models; a removable cord holder is included on corded models for easy storage. The cordless model can be used anywhere for up to 50 hours with an eight-hour charge; a charger is included.
Ordering information: This series includes a lifetime warranty.