Clear 360° Goniometer
Supplier: Fabrication Enterprises
12-1000
470019-280CS 788.05 CAD
470019-280
Clear 360° Goniometer
Goniometers
Clear Plastic Permits Easy Observation
This goniometer is marked in 1° increments and has three scales calibrated to be used with the International Standards of Measurement. One arm has a linear scale in both inches and centimeters to allow accurate measurement of the joint’s axis and range of motion.