1,6-Hexanediamine

Supplier: ALDON CORP SE

HH0023-100ML HH0026-100G
470300-004EA 25.6 CAD
470300-004 470300-006
1,6-Hexanediamine
Chemicals for Science Education

CAS Number: 124-09-4
Formula Weight: 116.21
Formula: 1,6-NH2(CH2)6NH2
Hazard Info: Corrosive
Density (g/mL): 0.854
Boiling Point (°C): 205
Freezing Point (°C): 39-42
Solubility: Water and Slightly in Alcohol
Synonyms: Hexamethyleneediamine, 1,6-Diaminohexane
Shelf Life (months): 36
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR